missing translation for 'onlineSavingsMsg'
Få mere at vide

Isopropyl 2-Furoate, TRC

Artikelnummer. 30179090
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30179090 250 mg -
30179677 500 mg -
30179222 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30179090 Leverandør Toronto Research Chemicals Leverandørnr. TRCI824605250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 6270-34-4
Molekylær formel C8H10O3
Molekylvægt (g/mol) 154.16
Synonym Isopropyl 2-furylcarboxylate, NSC 35576
IUPAC navn propan-2-yl furan-2-carboxylate
SMIL CC(C)OC(=O)c1occc1

Tekniske data

Kemisk navn eller materiale Isopropyl 2-Furoate
Mængde 250 mg
InChI formel InChI=1S/C8H10O3/c1-6(2)11-8(9)7-4-3-5-10-7/h3-6H,1-2H3
Formel vægt 154.063
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.