Få mere at vide
Isopropanol-d7, TRC
Artikelnummer. 30404040
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30404040 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404040 Leverandør Toronto Research Chemicals Leverandørnr. TRCI8227521G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isopropanol-d7
CAS 19214-96-1
Molekylær formel C3 2H7 H O
Mængde 1 g
Synonym 2-Propanol D7, Isopropyl-d7 alcohol (7CI,8CI), Isopropanol-d7, 2-Propan-1,1,1,2,3,3,3-d7-ol (9CI), 2-Propanol D7, Isopropyl-d7 alcohol (7CI,8CI), Isopropanol-d7, 2-Propan-1,1,1,2,3,3,3-d7-ol (9CI)
InChI formel InChI=1S/C3H8O/c1-3(2)4/h3-4H,1-2H3/i1D3,2D3,3D
SMIL [2H]C([2H])([2H])C([2H])(O)C([2H])([2H])[2H]
IUPAC navn 1,1,1,2,3,3,3-heptadeuteriopropan-2-ol
Molekylvægt (g/mol) 67.14
Formel vægt 67.1015