Learn More
Isoprene monoxide, 95%, Thermo Scientific Chemicals
Catalog Number p-7058518
Click to view available options
Quantity:
5 g
25 g
1838-94-4
C5H8O
84.118
MFCD00075225
FVCDMHWSPLRYAB-UHFFFAOYSA-N
2-methyl-2-vinyloxirane, isoprene monoxide, ccris 5685, 3,4-epoxy-3-methyl-1-butene, 2-methyl-2-vinyl-oxirane, oxirane, 2-ethenyl-2-methyl-, 2r, acmc-20m5m5, oxirane, 2-ethenyl-2-methyl, 4,4,4-3fmet-1-2-furyl-1,3-butandione
92166
2-ethenyl-2-methyloxirane
CC1(CO1)C=C
1838-94-4
C5H8O
84.118
MFCD00075225
FVCDMHWSPLRYAB-UHFFFAOYSA-N
2-methyl-2-vinyloxirane, isoprene monoxide, ccris 5685, 3,4-epoxy-3-methyl-1-butene, 2-methyl-2-vinyl-oxirane, oxirane, 2-ethenyl-2-methyl-, 2r, acmc-20m5m5, oxirane, 2-ethenyl-2-methyl, 4,4,4-3fmet-1-2-furyl-1,3-butandione
92166
2-ethenyl-2-methyloxirane
CC1(CO1)C=C
Isoprene monoxide is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isoprene monoxide is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. Recommended storage temperature is 2 - 8°C.
TRUSTED_SUSTAINABILITY
Density 0.857
Boiling Point 79°C to 81°C
Flash Point −8°C (17°F)
Odor Recognizable
Refractive Index 1.416
Quantity 5 g
UN Number UN1993
Beilstein 104216
Solubility Information Slightly soluble in water.
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material Isoprene monoxide