Få mere at vide
Isoprenaline Sulfate Dihydrate, Mikromol™
Beskrivelse
Isoprenaline Sulfate Dihydrate
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Isoprenaline Sulfate Dihydrate |
| CAS | 6078-56-4 |
| Analyt- eller komponentnavne | Isoprenaline Sulfate Dihydrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | 2 C11 H17 N O3 . 2 H2 O . H2 O4 S |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 1,2-Benzenediol, 4-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]-, sulfate, hydrate, 1,2-Benzenediol, 4-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]-, sulfate (2:1) (salt), dihydrate (9CI), Isoprenaline sulfate dihydrate, Isoproterenol sulfate dihydrate, dl-Isoproterenol sulfate dihydrate, 1,2-Benzenediol, 4-[1-hydroxy-2-[(1-methylethyl)amino]ethyl]-, (+/-)-, sulfate (2:1) (salt), dihydrate, Benzyl alcohol, 3,4-dihydroxy-alpha-[(isopropylamino)methyl]-, sulfate (2:1) (salt), dihydrate, (+/-)- |
| InChI formel | InChI=1S/2C11H17NO3.H2O4S.2H2O/c2*1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;1-5(2,3)4;;/h2*3-5,7,11-15H,6H2,1-2H3;(H2,1,2,3,4);2*1H2 |
| SMIL | O.O.CC(C)NCC(O)c1ccc(O)c(O)c1.CC(C)NCC(O)c2ccc(O)c(O)c2.OS(=O)(=O)O |
| IUPAC navn | 4-[1-hydroxy-2-(propan-2-ylamino)ethyl]benzene-1,2-diol;sulfuric acid;dihydrate |
| Vis mere |