Få mere at vide
Isopimaric Acid, TRC
Artikelnummer. 30135609
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135609 100 mg
17973445 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135609 Leverandør Toronto Research Chemicals Leverandørnr. TRCI821940100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5835-26-7
Molekylær formel C20 H30 O2
Molekylvægt (g/mol) 302.45
Synonym 1-Phenanthrenecarboxylic acid, 7-ethenyl-1,2,3,4,4a,4b,5,6,7,8,10,10a-dodecahydro-1,4a,7-trimethyl-, [1R-(1α,4aβ,4bα,7α,10aα)]-, Isopimaric acid (6CI), Podocarp-7-en-15-oic acid, 13β-methyl-13-vinyl- (7CI), Podocarp-7-en-15-oic acid, 13β-methyl-13-vinyl-, (-)- (8CI), (1R,4aR,4bS,7S,10aR)-7-Ethenyl-1,2,3,4,4a,4b,5,6,7,8,10,10a-dodecahydro-1,4a,7-trimethyl-1-phenanthrenecarboxylic acid, (+)-Isopimaric acid, 7,15-Isopimaradien-18-oic acid, Isopimaric acid A, δ7,15-Isopimaric acid
IUPAC navn (1R,4aR,4bS,7S,10aR)-7-ethenyl-1,4a,7-trimethyl-3,4,4b,5,6,8,10,10a-octahydro-2H-phenanthrene-1-carboxylic acid
SMIL C[C@@]1(CC[C@H]2C(=CC[C@@H]3[C@]2(C)CCC[C@@]3(C)C(=O)O)C1)C=C
Tekniske data
Kemisk navn eller materiale Isopimaric Acid
Mængde 100 mg
InChI formel InChI=1S/C20H30O2/c1-5-18(2)12-9-15-14(13-18)7-8-16-19(15,3)10-6-11-20(16,4)17(21)22/h5,7,15-16H,1,6,8-13H2,2-4H3,(H,21,22)/t15-,16+,18-,19+,20+/m0/s1
Formel vægt 302.2246