Få mere at vide
Isophthalic-2,4,5,6-d4 Acid, CDN
Artikelnummer. 30312444
Skift visning
Mængde:
0.5 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312444 0.5 g
30320287 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312444 Leverandør CDN Leverandørnr. CDND28190.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isophthalic Acid-2,4,5,6-D4
CAS 148472-48-4
Molekylær formel C8 D4 H2 O4
Mængde 0.5 g
Synonym 1,3-Benzene-2,4,5,6-d4-dicarboxylic acid (9 CI), Isophthalic Acid-d4, Isophthalic Acid-2,4,5,6-D4, Isophthalic-2,4,5,6-d4 Acid
InChI formel InChI=1 S/C8H6O4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4 H,(H,9,10)(H,11,12)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(C(=O)O)c([2 H])c(C(=O)O)c1[2 H]
IUPAC navn 2,4,5,6-tetradeuteriobenzene-1,3-dicarboxylic acid
Molekylvægt (g/mol) 170.1555
Formel vægt 170.0517 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity