Få mere at vide
Isophorone-d8 (2,4,4,6,6-d5; 3-methyl-d3), TRC
Artikelnummer. 30132732
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132732 5 mg
30132511 2.5 mg
17918649 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30132732 Leverandør Toronto Research Chemicals Leverandørnr. TRCI8241065MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 14397-59-2
Molekylær formel C9 D8 H6 O
Molekylvægt (g/mol) 146.26
Synonym Isophorone D8 (3-Methyl D3, 2,4,4,6,6 D5), 2-Cyclohexen-1-one-2,4,4,6,6-d5, 5,5-dimethyl-3-(methyl-d3)- (7CI,8CI,9CI)
IUPAC navn 2,4,4,6,6-pentadeuterio-5,5-dimethyl-3-(trideuteriomethyl)cyclohex-2-en-1-one
SMIL [2H]C1=C(C([2H])([2H])[2H])C([2H])([2H])C(C)(C)C([2H])([2H])C1=O
Tekniske data
Kemisk navn eller materiale Isophorone-d8 (2,4,4,6,6-d5; 3-methyl-d3)
Mængde 5 mg
InChI formel InChI=1S/C9H14O/c1-7-4-8(10)6-9(2,3)5-7/h4H,5-6H2,1-3H3/i1D3,4D,5D2,6D2
Formel vægt 146.1547