missing translation for 'onlineSavingsMsg'
Learn More
Isonicotinic acid, 99% Product Code.: 11409266

Isonicotinic acid, 99%

Product Code. p-7023994
100 g, 100g
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price

Product Code. 11409266

Brand: Thermo Scientific Alfa Aesar A18109.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price

CAS: 55-22-1 | C6H5NO2 | 123.111 g/mol

It acts as an useful isomer of nicotinic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It acts as an useful isomer of nicotinic acid.

Solubility
Soluble in water (6 mg/ml), and 2 N NH4OH (5%). Insoluble in ether, benzene, and boiling alcohol.

Notes
Store away from oxidizing agents and reducing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-22-1
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00006429
InChI Key TWBYWOBDOCUKOW-UHFFFAOYSA-N
Synonym isonicotinic acid, 4-pyridinecarboxylic acid, 4-picolinic acid, 4-carboxypyridine, p-pyridinecarboxylic acid, gamma-picolinic acid, gamma-pyridinecarboxylic acid, acide iso-nicotinique, 1,4-dihydroisonicotinic acid, acide iso-nicotinique french
PubChem CID 5922
ChEBI CHEBI:6032
IUPAC Name pyridine-4-carboxylic acid
SMILES C1=CN=CC=C1C(=O)O

Specifications

Melting Point ∼310°C (sublimation)
pH 3.0 to 4.0
Boiling Point 260°C (15 mmHg)
Quantity 100 g
Beilstein 109599
Merck Index 14,5187
Solubility Information Soluble in water (6mg/ml),and 2 N NH4OH (5%). Insoluble in ether,benzene,and boiling alcohol.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material Isonicotinic acid
compliance-icons
Product Identifier
  • Isonicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Isonicotinic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isonicotinic acid, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.