missing translation for 'onlineSavingsMsg'
Få mere at vide

Isonicotinamide-d4, TRC

Artikelnummer. 30405634
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30405634 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405634 Leverandør Toronto Research Chemicals Leverandørnr. TRCI82170210MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Isonicotinamide-2,3,5,6-d4
CAS 1219799-40-2
Molekylær formel C6 2H4 H2 N2 O
Mængde 10 mg
Synonym Isonicotinamide-d4, Isonicotinamide-2,3,5,6-D4, 4-Pyridinecarboxamide-2,3,5,6-d4, 4-(Aminocarbonyl)-2,3,5,6-tetradeuteriopyridine
InChI formel InChI=1S/C6H6N2O/c7-6(9)5-1-3-8-4-2-5/h1-4H,(H2,7,9)/i1D,2D,3D,4D
SMIL [2H]c1nc([2H])c([2H])c(C(=O)N)c1[2H]
IUPAC navn 2,3,5,6-tetradeuteriopyridine-4-carboxamide
Molekylvægt (g/mol) 126.15
Formel vægt 126.0731
Produkttitel
Vælg et problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.