Få mere at vide
Isonicotinamide-d4, TRC
Artikelnummer. 30405043
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30405043 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405043 Leverandør Toronto Research Chemicals Leverandørnr. TRCI821702100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isonicotinamide-2,3,5,6-d4
CAS 1219799-40-2
Molekylær formel C6 2H4 H2 N2 O
Mængde 100 mg
Synonym Isonicotinamide-d4, Isonicotinamide-2,3,5,6-D4, 4-Pyridinecarboxamide-2,3,5,6-d4, 4-(Aminocarbonyl)-2,3,5,6-tetradeuteriopyridine
InChI formel InChI=1S/C6H6N2O/c7-6(9)5-1-3-8-4-2-5/h1-4H,(H2,7,9)/i1D,2D,3D,4D
SMIL [2H]c1nc([2H])c([2H])c(C(=O)N)c1[2H]
IUPAC navn 2,3,5,6-tetradeuteriopyridine-4-carboxamide
Molekylvægt (g/mol) 126.15
Formel vægt 126.0731