Få mere at vide
Isometheptane Mucate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Isometheptene Mucate |
| CAS | 7492-31-1 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | 2 C9 H19 N . C6 H10 O8 |
| Mængde | 25 mg |
| Synonym | Galactaric acid, compd. with N,6-dimethyl-5-hepten-2-amine (1:2) (9CI), Galactaric acid, compd. with N,1,5-trimethyl-4-hexenylamine (1:2) (8CI), Mucic acid, salt with N,1,5-trimethyl-4-hexenylamine (1:2) (6CI), 4-Hexenylamine, N,1,5-trimethyl-, galactarate (2:1) (8CI), 5-Hepten-2-amine, N,6-dimethyl-, galactarate (2:1) (9CI), Isometheptene mucate |
| InChI formel | InChI=1S/2C9H19N.C6H10O8/c2*1-8(2)6-5-7-9(3)10-4;7-1(3(9)5(11)12)2(8)4(10)6(13)14/h2*6,9-10H,5,7H2,1-4H3;1-4,7-10H,(H,11,12)(H,13,14)/t;;1-,2+,3+,4- |
| SMIL | CNC(C)CCC=C(C)C.CNC(C)CCC=C(C)C.O[C@H]([C@H](O)[C@@H](O)C(=O)O)[C@H](O)C(=O)O |
| IUPAC navn | N,6-dimethylhept-5-en-2-amine;(2R,3S,4R,5S)-2,3,4,5-tetrahydroxyhexanedioic acid |
| Molekylvægt (g/mol) | 492.65 |
| Vis mere |