Få mere at vide
Isometamidium chloride hydrochloride 1000 μg/mL in Acetonitrile:Water, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Isometamidium Chloride Hydrochloride |
| CAS | 6798-24-9 |
| Analyt- eller komponentnavne | Isometamidium Chloride Hydrochloride |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C28 H26 N7 . Cl . Cl H |
| Koncentration | 1000 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| Synonym | Phenanthridinium, 3-amino-8-[3-[3-(aminoiminomethyl)phenyl]-2-triazen-1-yl]-5-ethyl-6-phenyl-, chloride, hydrochloride (1:1:1), 8-[3-(m-Amidinophenyl)-2-triazeno]-3-amino-5-ethyl-6-phenylphenanthridinium chloride, hydrochloride (7CI), Isometamidium hydrochloride (6CI), Phenanthridinium, 3-amino-8-[3-[3-(aminoiminomethyl)phenyl]-1-triazenyl]-5-ethyl-6-phenyl-, chloride, monohydrochloride (9CI), Phenanthridinium, 8-[3-(m-amidinophenyl)-2-triazeno]-3-amino-5-ethyl-6-phenyl-, chloride, monohydrochloride (8CI), Isometamidium chloride hydrochloride, Samorin, Trypamidium, veridium |
| InChI formel | InChI=1S/C28H25N7.2ClH/c1-2-35-26-16-20(29)11-13-24(26)23-14-12-22(17-25(23)27(35)18-7-4-3-5-8-18)33-34-32-21-10-6-9-19(15-21)28(30)31;;/h3-17,29H,2H2,1H3,(H4,30,31,32,33);2*1H |
| SMIL | Cl.[Cl-].CC[n+]1c(c2ccccc2)c3cc(ccc3c4ccc(N)cc14)N=NNc5cccc(c5)C(=N)N |
| Vis mere |