missing translation for 'onlineSavingsMsg'
Få mere at vide

(+)-Isomenthone, TRC

Artikelnummer. 17974755
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17974755 100 mg
17144901 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17974755 Leverandør Toronto Research Chemicals Leverandørnr. TRCM199545100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1196-31-2
Molekylær formel C10 H18 O
Molekylvægt (g/mol) 154.25
Synonym (+)-Isomenthone, (2R,5R)-2-Isopropyl-5-methyl-cyclohexanone, (1R,4R)-Isomenthone, (1R,4R)-p-Menth-3-one, D-Isomenthone, L-Isomenthone, d-Isomenthone, d-Menthone
IUPAC navn (2R,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one
SMIL CC(C)[C@H]1CC[C@@H](C)CC1=O

Specifications

Kemisk navn eller materiale Isomenthone
Renhedsgrad noter GC
Mængde 100 mg
InChI formel InChI=1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3/t8-,9-/m1/s1
Formel vægt 154.1358
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.