Få mere at vide
(±)-Isomenthone (Contains ~20% of trans-isomer), TRC
Artikelnummer. 30137075
Skift visning
Mængde:
250 mg
2.5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137075 2.5 g
17944885 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137075 Leverandør Toronto Research Chemicals Leverandørnr. TRCI8213302.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 491-07-6
Molekylær formel C10 H18 O
Molekylvægt (g/mol) 154.25
Synonym Cyclohexanone, 5-methyl-2-(1-methylethyl)-, (2R,5R)-rel-, Cyclohexanone, 5-methyl-2-(1-methylethyl)-, cis-, p-Menthan-3-one, cis- (8CI), (+/-)-Isomenthone, Isomenthone, cis-Menthone, cis-p-Menthan-3-one, cis-p-Menthone, dl-Isomenthone, rel-(2R,5R)-5-Methyl-2-(1-methylethyl)cyclohexanone, alpha-Isomenthone
IUPAC navn (2R,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one
SMIL CC(C)[C@H]1CC[C@@H](C)CC1=O
Tekniske data
Kemisk navn eller materiale (±)-Isomenthone
Mængde 2.5 g
InChI formel InChI=1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3/t8-,9-/m1/s1
Formel vægt 154.1358