Få mere at vide
Isoleucine-d10 (mixture of diastereomers), CDN
Artikelnummer. 30320105
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320105 0.05 g
30324300 0.1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320105 Leverandør CDN Leverandørnr. CDND79300.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isoleucine-d10 (mixture of diastereomers)
CAS 29909-02-2
Molekylær formel C6 D10 H3 N O2
Mængde 0.05 g
Synonym Isoleucine-2,3,3',3',3',4,4,5,5,5-d10 (9 CI), Isoleucine-2,3,4,4,4',4',4',5,5,5-d10 (8 CI)
InChI formel InChI=1 S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/i1D3,2D3,3D2,4 D,5 D
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])(N)C(=O)O
IUPAC navn 2-amino-2,3,4,4,5,5,5-heptadeuterio-3-(trideuteriomethyl)pentanoic acid
Molekylvægt (g/mol) 141.2345
Formel vægt 141.1574 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity