Få mere at vide
Isohexanol-d7, TRC
Artikelnummer. 30404910
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30404910 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404910 Leverandør Toronto Research Chemicals Leverandørnr. TRCI819617250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isohexanol-d7
CAS 1246819-30-6
Molekylær formel C6H7D7O
Mængde 250 mg
Synonym 4-Methyl-1-pentanol-d7, Anglamol 6085U-d7, NSC 91492-d7, iso-Hexanol-d7
InChI formel InChI=1S/C6H14O/c1-6(2)4-3-5-7/h6-7H,3-5H2,1-2H3/i1D3,2D3,6D
SMIL OCCCC([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 4-(methyl-d3)pentan-4,5,5,5-d4-1-ol
Molekylvægt (g/mol) 109.22
Formel vægt 109.15