missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Isoguvacine hydrochloride, MedChemExpress
Description
Isoguvacine hydrochloride is a GABA receptor agonist.
Specifications
Specifications
| Kemisk navn eller materiale | Isoguvacine hydrochloride |
| CAS | 68547-97-7 |
| Farve | Yellow |
| Renhedsgrad noter | Research |
| Molekylær formel | C6H10ClNO2 |
| Mængde | 100 mg |
| Opløselighedsinformation | H2O : 100 mg/mL (611.25 mM; Need ultrasonic) ∣DMSO : 25 mg/mL (152.81 mM; Need ultrasonic) |
| SMIL | O=C(C1=CCNCC1)O.[H]Cl |
| Molekylvægt (g/mol) | 163.6 |
| Formel vægt | 163.6 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?