missing translation for 'onlineSavingsMsg'
Få mere at vide

Isoflavone, TRC

Artikelnummer. 30390457
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30390457 25 mg
30406460 250 mg
30399391 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30390457 Leverandør Toronto Research Chemicals Leverandørnr. TRCI81680825MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 574-12-9
Molekylær formel C15H10O2
Molekylvægt (g/mol) 222.24
Synonym 3-Phenyl-4H-1-benzopyran-4-one, 3-Phenyl-4-chromone, 3-Phenylchromone, NSC 135405, Isoflavone 80
SMIL O=C1C(=COc2ccccc12)c3ccccc3

Specifications

Kemisk navn eller materiale Isoflavone
Mængde 25 mg
InChI formel InChI=1S/C15H10O2/c16-15-12-8-4-5-9-14(12)17-10-13(15)11-6-2-1-3-7-11/h1-10H
Formel vægt 222.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.