missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Isocytosine, 99%

Catalog Number p-7035992
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
108-53-2
C4H5N3O
111.10
MFCD00057557 MFCD00023256
XQCZBXHVTFVIFE-UHFFFAOYSA-N
isocytosine, 2-amino-4-hydroxypyrimidine, 2-aminopyrimidin-4-ol, 2-aminopyrimidin-4 1h-one, 2-amino-4-pyrimdinol, 2-amino-4-pyrimidone, 2-amino-4-oxypyrimidine, 2-aminouracil, 4-pyrimidinol, 2-amino, 4 1h-pyrimidinone, 2-amino
66950
CHEBI:55502
2-amino-3,4-dihydropyrimidin-4-one
NC1=NC=CC(=O)N1
This item is not returnable. View return policy
108-53-2
C4H5N3O
111.10
MFCD00057557 MFCD00023256
XQCZBXHVTFVIFE-UHFFFAOYSA-N
isocytosine, 2-amino-4-hydroxypyrimidine, 2-aminopyrimidin-4-ol, 2-aminopyrimidin-4 1h-one, 2-amino-4-pyrimdinol, 2-amino-4-pyrimidone, 2-amino-4-oxypyrimidine, 2-aminouracil, 4-pyrimidinol, 2-amino, 4 1h-pyrimidinone, 2-amino
66950
CHEBI:55502
2-amino-3,4-dihydropyrimidin-4-one
NC1=NC=CC(=O)N1

Isocytosine (2-aminouracil) is an isomer of cytosine used in physical chemical studies involving metal complex binding, hydrogen-bonding, and tautomerism and proton transfer effects in nucleobases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isocytosine (2-aminouracil) is an isomer of cytosine used in physical chemical studies involving metal complex binding, hydrogen-bonding, and tautomerism and proton transfer effects in nucleobases.

Solubility
Soluble in DMF, DMSO, hot water, and AcOH.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Soluble in DMF,DMSO,hot water,and AcOH.
Formula Weight 111.1
Percent Purity 99%
Chemical Name or Material Isocytosine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'