Få mere at vide
Isobuzole, LoGiCal
Artikelnummer. 30464234
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30464234 5 mg
1 options
Artikelnummer. 30464234 Leverandør LoGiCal Leverandørnr. LGCFOR3113.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isobuzole
CAS 3567-08-6
Analyt- eller komponentnavne Isobuzole
Forsendelsestilstand Room Temperature
Molekylær formel C13 H17 N3 O3 S2
Certificeringer/compliance ISO 9001
Synonym Benzenesulfonamide, 4-methoxy-N-[5-(2-methylpropyl)-1,3,4-thiadiazol-2-yl]-, Benzenesulfonamide, N-(5-isobutyl-1,3,4-thiadiazol-2-yl)-p-methoxy- (6CI,7CI,8CI), 2-(p-Methoxybenzenesulfonamido)-5-isobutyl-1,3,4-thiadiazole, 8002 C. B., Glysobuzole, Isobuzol, Isobuzole, N-(5-Isobutyl-1,3,4-thiadiazol-2-yl)-p-methoxybenzenesulfonamide, NSC 34014, Stabinol
InChI formel InChI=1S/C13H17N3O3S2/c1-9(2)8-12-14-15-13(20-12)16-21(17,18)11-6-4-10(19-3)5-7-11/h4-7,9H,8H2,1-3H3,(H,15,16)
Løsningstype Neat
SMIL COc1ccc(cc1)S(=O)(=O)Nc2nnc(CC(C)C)s2
IUPAC navn 4-methoxy-N-[5-(2-methylpropyl)-1,3,4-thiadiazol-2-yl]benzenesulfonamide
Industry Type Clinical Testing
Molekylvægt (g/mol) 327.42
Mængde 5 mg
Formel vægt 327.0711 g/mol