Få mere at vide
Isobutyl Thiuram Disulfide, TRC
Artikelnummer. 30405353
Skift visning
Mængde:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30405353 250 mg
30390919 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30405353 Leverandør Toronto Research Chemicals Leverandørnr. TRCI788750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isobutyl Thiuram Disulfide
CAS 3064-73-1
Molekylær formel C18 H36 N2 S4
Mængde 250 mg
InChI formel InChI=1S/C18H36N2S4/c1-13(2)9-19(10-14(3)4)17(21)23-24-18(22)20(11-15(5)6)12-16(7)8/h13-16H,9-12H2,1-8H3
SMIL CC(C)CN(CC(C)C)C(=S)SSC(=S)N(CC(C)C)CC(C)C
IUPAC navn bis(2-methylpropyl)carbamothioylsulfanyl N,N-bis(2-methylpropyl)carbamodithioate
Molekylvægt (g/mol) 408.752
Formel vægt 408.176