Få mere at vide
Isobutyl Methyl Ketone, Supelco™
Beskrivelse
EMSURE grade solvents are suitable for a broad spectrum of classical lab applications, and are frequently used in regulated and highly demanding lab applications. EMSURE provides worldwide best and most extensive product specifications. We declare our EMSURE range to be in compliance with the ACS, with the reagent part of the European Pharmacopoeia (Reag. Ph Eur) and also with the ISO standards.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Isobutyl Methyl Ketone |
| Smeltepunkt | −80°C (lit.) |
| Tæthed | 0.801 g/mL at 25°C |
| Mængde | 2.5 L |
| Molekylær formel | (CH3)2CHCH2COCH3 |
| MDL nummer | MFCD00008938 |
| Synonym | 4-Methyl-2-pentanone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone, 4-Methyl-2-pentanone, Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone |
| Kogepunkt | 117°C to 118°C |
| Molekylvægt (g/mol) | 100.16 |
| Grad | Research |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.