Få mere at vide
Isobutyl-D9 Paraben, TRC
Artikelnummer. 30404917
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30404917 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30404917 Leverandør Toronto Research Chemicals Leverandørnr. TRCI7801025MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isobutyl-d9 Paraben
CAS 1215460-35-7
Molekylær formel C11H5D9O3
Mængde 5 mg
Synonym 4-Hydroxybenzoic Acid 2-Methylpropyl-d9 Ester, 2-Methylpropyl-d9 p-Hydroxybenzoate, Isobutyl-d9 4-Hydroxybenzoate, Isobutyl-d9 p-Hydroxybenzoate, p-Hydroxybenzoic Acid Isobutyl-d9 Ester
InChI formel InChI=1S/C11H14O3/c1-8(2)7-14-11(13)9-3-5-10(12)6-4-9/h3-6,8,12H,7H2,1-2H3/i1D3,2D3,7D2,8D
SMIL OC1=CC=C(C(OC([2H])([2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)C=C1
Molekylvægt (g/mol) 203.28
Formel vægt 203.1508