Learn More
Isobenzofuran-1(3H)-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 87-41-2 |
| Molecular Formula | C8H6O2 |
| MDL Number | MFCD00005906 |
| Quantity | 10 g |
| SMILES | O=C1OCC2=C1C=CC=C2 |
| Molecular Weight (g/mol) | 134.13 |
| Percent Purity | 98% |
| Chemical Name or Material | Isobenzofuran-1(3H)-one |