Få mere at vide
Isoamyl Benzoate, LoGiCal
Artikelnummer. 30464073
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464073 10 mg
1 options
Artikelnummer. 30464073 Leverandør LoGiCal Leverandørnr. LGCFOR1086.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isoamyl Benzoate
CAS 94-46-2
Analyt- eller komponentnavne Isoamyl Benzoate
Forsendelsestilstand Room Temperature
Molekylær formel C12 H16 O2
Certificeringer/compliance ISO 9001
Synonym Isoamyl Benzoate, 3-Methyl-1-butanol 1-benzoate, 3-Methyl-1-butanol benzoate, Isopentyl alcohol benzoate, 3-Methylbutyl benzoate, Isoamyl benzoate, Isopentyl benzoate, NSC 9284, iso-Amyl benzoate, iso-Pentyl benzoate
InChI formel InChI=1S/C12H16O2/c1-10(2)8-9-14-12(13)11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3
Løsningstype Neat
SMIL CC(C)CCOC(=O)c1ccccc1
IUPAC navn 3-methylbutyl benzoate
Industry Type Clinical Testing
Molekylvægt (g/mol) 192.25
Mængde 10 mg
Formel vægt 192.115 g/mol