Få mere at vide
Iso Sildenafil-d3, TRC
Artikelnummer. 30407418
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30407418 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407418 Leverandør Toronto Research Chemicals Leverandørnr. TRCI90080210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Iso Sildenafil-d3
CAS 1794829-17-6
Molekylær formel C22H27D3N6O4S
Mængde 10 mg
Synonym 5-[2-Ethoxy-5-[(4-(methyl-d3)-1-piperazinyl)sulfonyl]phenyl]-2,6-dihydro-2-methyl-3-propyl-7H-pyrazolo[4,3-d]pyrimidin-7-one, 1-[[3-(4,7-Dihydro-2-methyl-7-oxo-3-propyl-2H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-(methyl-d3)piperazine
InChI formel InChI=1S/C22H30N6O4S/c1-5-7-17-19-20(25-27(17)4)22(29)24-21(23-19)16-14-15(8-9-18(16)32-6-2)33(30,31)28-12-10-26(3)11-13-28/h8-9,14H,5-7,10-13H2,1-4H3,(H,23,24,29)/i3D3
SMIL [2H]C([2H])([2H])N1CCN(CC1)S(=O)(=O)c2ccc(OCC)c(c2)C3=NC(=O)c4nn(C)c(CCC)c4N3
IUPAC navn 5-[2-ethoxy-5-[4-(trideuteriomethyl)piperazin-1-yl]sulfonylphenyl]-2-methyl-3-propyl-4H-pyrazolo[4,3-d]pyrimidin-7-one
Molekylvægt (g/mol) 477.59
Formel vægt 477.2238