Få mere at vide
Brady™ ISO Safety Sign - Disinfect Your Hands, Universal
Artikelnummer. 30280476
Skift visning
Materiale:
Aluminium
Polyester
Polypropylen
Diameter (metrisk):
150 mm
200 mm
315 mm
400 mm
500 mm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Materiale Diameter (metrisk)
30280476 Polyester 500 mm
30280545 Polypropylen 400 mm
30280495 Aluminium 150 mm
30280533 Aluminium 200 mm
30280531 Aluminium 315 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 30280476 Leverandør Brady™ Leverandørnr. 322319
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Sign to signify that hands must be disinfected.
  • Sign compliant to ISO 7010
Tekniske data
Certificeringer/compliance ISO 7010; Compliant with Directive 2011/65/EU and 2015/863
Alternativt varenummer M/M061/NT/FLO-DIA500-1
Grad M061
Materiale Polyester
Mængde 1 Ea.
Diameter (metrisk) 500 mm
Monteringstype Surface Mount, Wall Mount
Til brug med (applikation) Floor Marking
Tykkelse 0.28 mm
Form Round
Sprog Universal
Vist tekst Disinfect your hands
Produkttype Personal Hygiene Sign
Farve White on Blue