Få mere at vide
iso-Propyl-1,1,1,3,3,3-d6-amine HCl, CDN
Artikelnummer. 30316807
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30316807 0.5 g 0.5g
30325911 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316807 Leverandør CDN Leverandørnr. CDND56860.5G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Propanamine-1,1,1,3,3,3-D6 Hydrochloride
CAS 126794-59-0
Molekylær formel C3 D6 H3 N . Cl H
Mængde 0.5 g
Synonym 2-Propan-1,1,1,3,3,3-d6-amine, hydrochloride (9 CI), Isopropyl-d6-amine hydrochloride, Isopropyl-1,1,1,3,3,3-d6-amine hydrochloride, 2-Aminopropane-1,1,1,3,3,3-d6 hydrochloride, 2-Aminopropane-1,1,1,3,3,3-D6 hydrochloride, 2-Propanamine-1,1,1,3,3,3-D6 hydrochloride
InChI formel InChI=1 S/C3H9N.ClH/c1-3(2)4;/h3H,4H2,1-2H3;1 H/i1D3,2D3;
SMIL Cl.[2 H]C([2 H])([2 H])C(N)C([2 H])([2 H])[2 H]
IUPAC navn 1,1,1,3,3,3-hexadeuteriopropan-2-amine;hydrochloride
Molekylvægt (g/mol) 101.6082
Formel vægt 101.0878 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity