Få mere at vide
iso-Colchicine-d3, TRC
Artikelnummer. 30368217
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30368217 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30368217 Leverandør Toronto Research Chemicals Leverandørnr. TRCC64000650MG
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale iso-Colchicine-d3
CAS 1246818-69-8
Molekylær formel C22H22D3NO6
Mængde 50 mg
Synonym N-[(7S)-5,6,7,10-Tetrahydro-1,2,3,9-tetramethoxy-10-oxobenzo[a]heptalen-7-yl]acetamide, Isocolchicine
InChI formel InChI=1S/C22H25NO6/c1-12(24)23-16-8-6-13-10-19(27-3)21(28-4)22(29-5)20(13)14-7-9-17(25)18(26-2)11-15(14)16/h7,9-11,16H,6,8H2,1-5H3,(H,23,24)/t16-/m0/s1/i1D3
SMIL O=C(C([2H])([2H])[2H])N[C@@H](CC1)C2=C(C=CC(C(OC)=C2)=O)C3=C1C=C(OC)C(OC)=C3OC
IUPAC navn 2,2,2-trideuterio-N-[(7S)-1,2,3,9-tetramethoxy-10-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl]acetamide
Molekylvægt (g/mol) 402.46