Få mere at vide
iso-Butyl 4-Hydroxybenzoate-2,3,5,6-d4, TRC
Artikelnummer. 30407611
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30407611 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407611 Leverandør Toronto Research Chemicals Leverandørnr. TRCI7801011MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxybenzoic acid-isobutyl ester ring-d4
CAS 1219805-33-0
Molekylær formel C11 2H4 H10 O3
Mængde 1 mg
Synonym 4-Hydroxybenzoic acid-isobutyl ester D4 (ring D4)
InChI formel InChI=1S/C11H14O3/c1-8(2)7-14-11(13)9-3-5-10(12)6-4-9/h3-6,8,12H,7H2,1-2H3/i3D,4D,5D,6D
SMIL [2H]c1c([2H])c(C(=O)OCC(C)C)c([2H])c([2H])c1O
IUPAC navn 2-methylpropyl 2,3,5,6-tetradeuterio-4-hydroxybenzoate
Molekylvægt (g/mol) 198.25
Formel vægt 198.1194