Få mere at vide
STEMCELL Technologies ISCK03
Beskrivelse
- ISCK03 (inhibitor of SCF/c-KIT) is a cell-permeable phenyl-imidazolosulfonamide compound that inhibits KIT activity
- ISCK03 inhibits SCF-induced c-KIT phosphorylation and downstream ERK phosphorylation at a concentration of 1 - 5 μM in 501mel melanoma cells (Na et al.)
- Causes significant decrease in blood vessel density on human lung adenocarcinoma (A549) cells irradiated with X-rays (6 Gy; Kamlah et al.)
- Reduces cell viability and ERK1/2 phosphorylation in acute myeloid leukemia (AML)-derived cell lines (Kon et al.)
Tekniske data
Tekniske data
| CAS | 945526-43-2 |
| Synonym | c-Kit inhibitor II; Stem cell factor/c-Kit inhibitor |
| Mængde | 10 mg |
| Molekylær formel | C19H21N3O2S |
| Renhed | ≥95% |