Få mere at vide
Isavuconazole-d4, TRC
Artikelnummer. 30403767
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30403767 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30403767 Leverandør Toronto Research Chemicals Leverandørnr. TRCI7778321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Isavuconazole-d4
CAS 1346598-58-0
Renhedsgrad noter HPLC
Molekylær formel C22H13D4F2N5OS
Mængde 1 mg
Synonym 4-[2-[(1R,2R)-2-(2,5-Difluorophenyl)-2-hydroxy-1-methyl-3-(1H-1,2,4-triazol-1-yl)propyl]-4-thiazolyl]benzonitrile-d4, 2R,3R)-3-[4-(4-Cyanophenyl)thiazol-2-yl]-2-(2,5- difluorophenyl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol-d4, BAL 4815-d4, RO 0094815-d4
InChI formel InChI=1S/C22H17F2N5OS/c1-14(21-28-20(10-31-21)16-4-2-15(9-25)3-5-16)22(30,11-29-13-26-12-27-29)18-8-17(23)6-7-19(18)24/h2-8,10,12-14,30H,11H2,1H3/t14-,22+/m0/s1/i2D,3D,4D,5D
SMIL FC1=CC([C@](CN2C=NC=N2)(O)[C@@H](C)C3=NC(C4=C([2H])C([2H])=C(C#N)C([2H])=C4[2H])=CS3)=C(F)C=C1
Molekylvægt (g/mol) 441.49
Formel vægt 441.1373
Procent renhed >95