Learn More
Chemsplash Irradiated Zip Labcoat PB(6B) Elastic Cuff
GREENER_CHOICE
Supplied Gamma Sterile Irradiated and double bagged. This lab coat is a 2/3 length coat with a collar and zip closure and elasticated wrists.
Brand:  Chemsplash 2635.400.009
 View more versions of this product
Product Code. 30058289
  • 2920.00 DKK / Pack of 50
Only null left
Estimated Shipment: 11-03-2026
Add to basket
This item is not returnable. View return policy
Description
Description
Chemsplash Sterile Lab Coat
  • Chemsplash 55GSM Type 5B and 6B PB Fabric
  • GAMMA Irradiated and Double Bagged
  • Zip Fasten Front with Adhesive Flap Cover
  • Elasticated Cuffs
  • Fold Over Collar
  • Silicone and Latex Free
  • Anti-Static Material
  • Non-Linting
  • Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: Supplied GAMMA Sterile Irradiated and double bagged, the Chemsplash Lab Coat is made from Chemsplash fabric, which is soft and microporous. It is strong and resistant to chemical splash and dust. This lab coat is a 2/3 length coat with a collar and zip closure and elasticated wrists. It is anti-static to meet with EN1149-5:2008, lint-free and suitable for clean room applications. Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9. Chemsplash accessories can be used to protect the wearer from the environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifications
Specifications
Lab Coat
Unisex
White
Elastic
Sterile
None
Autoclavable
3X-Large
Microporous Composite Fabric
Zipper
Single-use
Liquids
Yes
Class 100-100,000 (ISO 5-8)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Chemsplash Irradiated Zip Labcoat PB(6B) Elastic Cuff > 3XL