missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Iron Saccharate, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Iron Saccharate |
| CAS | 8047-67-4 |
| Molekylær formel | [Na2Fe5O8(OH) • 3H2O]n •(C12H22O11)m |
| Mængde | 25 g |
| Synonym | Colliron IV, Feojectin, Ferric Hydroxide Sucrose Complex, Ferrivenin, Ferrum Hausmann, Ferrum Vitis, Ferum Hausmann, Fesin, Hippiron, Iron Oxide Saccharated, Iron Sucrose, Iron Sucrose Complex, Iron Sugar, Iron(III) Hydroxide Sucrose Complex, Iviron, Neoferrum, Proferrin, Saccharated Ferric Oxide, Saccharated Iron, Saccharated Iron Oxide, Sucrofer, Venofer, Venoferrum, XI 921 |
| InChI formel | InChI=1S/3C6H10O8.2Fe/c3*7-1(3(9)5(11)12)2(8)4(10)6(13)14;;/h3*1-4,7-10H,(H,11,12)(H,13,14);;/q;;;2*+3/p-6/t3*1-,2-,3-,4+;;/m000../s1 |
| SMIL | [C@H]([C@@H]([C@@H](C(=O)[O-])O)O)([C@H](C(=O)[O-])O)O.[C@H]([C@@H]([C@@H](C(=O)[O-])O)O)([C@H](C(=O)[O-])O)O.[C@H]([C@@H]([C@@H](C(=O)[O-])O)O)([C@H](C(=O)[O-])O)O.[Fe+3].[Fe+3] |
| Molekylvægt (g/mol) | 736.1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?