missing translation for 'onlineSavingsMsg'
Learn More

Iron, plasma standard solution, Specpure™ Fe 10,000μg/mL, Thermo Scientific Chemicals

Catalog Number 15427485
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy
MFCD00010999
This item is not returnable. View return policy
MFCD00010999

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 50 mL
UN Number UN3265
Grade Specpure™
Odor Acidic
Physical Form Liquid
Chemical Name or Material Iron, plasma standard solution, Fe 10,000μg/mL
compliance-icons
Product Identifier
  • Iron, plasma standard solution, Specpure, Fe 10,000µg/ml
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : Iron (III) nitrate nonahydrate, Nitric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'