missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) oxide, 99.85+% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
It has many applications, mainly used in iron industry, pigment, as photocatalyst, medicine and polishing jewels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 1565°C |
Color | Red to Brown |
Physical Form | Sintered Lump |
Quantity | 50 g |
Assay Percent Range | >99.85% (Metals basis) |
Solubility Information | Insoluble in water,Soluble in diluted acids,sugar solution and slightly soluble in aq. tartaric acid,citric acid,CH3COOH. |
Formula Weight | 159.69 |
Odor | Odorless |
Density | 5.24 |
Chemical Name or Material | Iron(III) oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'