missing translation for 'onlineSavingsMsg'
Learn More

Iron(III) nitrate nonahydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals

Catalog Number p-4407482
Click to view available options
Quantity:
500 g
2 kg
5 x 2 kg
7782-61-8
FeH18N3O18
403.99
MFCD00149708
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
16211566
iron(3+) nonahydrate trinitrate
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
7782-61-8
FeH18N3O18
403.99
MFCD00149708
SZQUEWJRBJDHSM-UHFFFAOYSA-N
ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
16211566
iron(3+) nonahydrate trinitrate
O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

It is used as catalyst for the synthesis of sodium amide from sodium and ammonia, and for etching of silver and silver alloys. Ferric nitrate is used in the synthesis of organically templated iron phosphates, and acts as an efficient catalyst in enamination of beta dicarbonyl compounds to beta enaminones. Ferric nitrate impregnated clays are useful as oxidizing agents. Clayfen, produced from ferric nitrate and montmorillonite, is used for the oxidation of thiols to disulfide, and alcohols to aldehydes. Ferric nitrate is useful for regio- and stereo-selective nitration of various aromatic, aliphatic, and heteroaromatic olefins. This reaction provides nitroolefins with excellent E-selectivity. With Keggin-type heteropoly acids, it selectively oxidizes various hydroxy groups.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 125°C (decomposition)
Melting Point 47°C
Color Purple
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 98.0 to 101.0%
Solubility Information Freely soluble in water,alcohol,acetone. Slightly soluble in cold and concentrated HNO3Soluble in water,alcohol and acetone.
Formula Weight 403.99 (241.86 Anhydrous)
Grade ACS
Odor Acidic
Sensitivity Moisture sensitive
Appearance Pale purple
Density 1.684 g/cm3
Chemical Name or Material Iron(III) nitrate nonahydrate, For ACS analysis
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(III) nitrate nonahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'