missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) meso-tetraphenylporphine chloride, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
Iron(III) meso-tetraphenylporphine chloride is used catalyzing reagent for silylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Crystalline |
Quantity | 250 mg |
Solubility Information | Insoluble in water. |
Formula Weight | 704.04 |
Chemical Name or Material | Iron(III) meso-tetraphenylporphine chloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'