missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) hydroxide, alpha-phase, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Iron(III) hydroxide is used in purifying water and as an absorbent in chemical processes. It is also used as a pigment and as a catalyst. Further, it is used in some tattoo ink and in aquarium water treatment as a phosphate binder.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown |
Physical Form | Powder |
Quantity | 5 g |
Solubility Information | Soluble in mineral acids. Insoluble in water and ethanol. |
Formula Weight | 88.85 |
Appearance | Brown powder |
Density | 3.4 to 3.9 g/cm3 |
Chemical Name or Material | Iron(III) hydroxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'