missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) ethoxide, soluble polymeric mixture, Thermo Scientific™
Click to view available options
Quantity:
2 g
10 g
50 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | Brown |
Boiling Point | (decomposition) |
Quantity | 2 g |
UN Number | UN2926 |
Solubility Information | Soluble in hexane and ethanol |
Physical Form | Solid |
Chemical Name or Material | Iron (III) ethoxide, Soluble polymeric mixture |
Safety and Handling
Product Identifier
- Iron(III) ethoxide, soluble polymeric mixture
- Danger
- Flammable solid Category 2
- Skin corrosion/irritation Category 1 B
- H228-Flammable solid.
- H314-Causes severe skin burns and eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'