missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(III) citrate hydrate, Fe(III) 16.5-20%; Fe(II) max 5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1000 g
Description
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.
Solubility
Soluble in water.
Notes
Light sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Provides a source of iron used in cell culture applications. Provides iron in a less toxic form than free iron salts.
Solubility
Soluble in water.
Notes
Light sensitive. Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Quantity | 50 g |
Solubility Information | Soluble in water. |
Formula Weight | 244.94 (Anhydrous) |
Concentration or Composition (by Analyte or Components) | Fe(III) 16.5 to 20%; Fe(II) max 5% |
Sensitivity | Light sensitive |
Chemical Name or Material | Iron (III) citrate hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'