missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Iron(III) chloride, Sigma-Aldrich™
Beskrivelse
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Iron(III) chloride |
| Smeltepunkt | 306°C (decomposition) |
| CAS | 7705-08-0 |
| Mængde | 25 kg |
| Tæthed | 2.89 g/cm³ at 25°C |
| Synonym | Iron(III) chloride, Ferric chloride, Iron trichloride, Iron(III) chloride |
| Molekylvægt (g/mol) | 162.2 |
| Grad | Not Available |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?