Learn More
Iron(III) chloride, MilliporeSigma™
Product Code. 15886370
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
Product Code. Quantity Unit Size
15886370 500 g 500g
1 options
Product Code. 15886370 Supplier Sigma Aldrich Supplier No. 8039450500
 Restricted Item
Forventet forsendelse: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Specifications
CAS 7705-08-0
Physical Form Powder
Quantity 500 g
Odor Stinging
Vapor Pressure 1hPa (20°C)
Density 2.89g/cm3 at 25°C
Molecular Formula Cl3Fe
MDL Number MFCD00011005
UN Number UN1773
Synonym Ferric chloride; Iron trichloride
Solubility Information Soluble in water: 920g/L at 25°C
InChI Key RBTARNINKXHZNM-UHFFFAOYSA-K
SMILES [Cl-].[Cl-].[Cl-].[Fe+3]
IUPAC Name iron(3+) trichloride
Molecular Weight (g/mol) 162.20
Formula Weight 162.2
Percent Purity ≥98.0%
Grade Not Available
Chemical Name or Material Iron(III) chloride