Få mere at vide
Iron(II) Trifluoromethanesulfonate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Iron(II) Trifluoromethanesulfonate |
| CAS | 59163-91-6 |
| Molekylær formel | C2F6FeO6S2 |
| Mængde | 250 mg |
| Synonym | Iron Bis(trifluoromethanesulfonate), Iron Ditriflate, Iron(2+) Triflate, Iron(2+) Trifluoromethanesulfonate, Iron(II) Triflate, Iron(II) Trifluoromethanesulfonate |
| InChI formel | InChI=1S/2CHF3O3S.Fe/c2*2-1(3,4)8(5,6)7;/h2*(H,5,6,7);/q;;+2/p-2 |
| SMIL | [Fe+2].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
| IUPAC navn | iron(2+);trifluoromethanesulfonate |
| Molekylvægt (g/mol) | 353.98 |
| Formel vægt | 353.839 |