missing translation for 'onlineSavingsMsg'
Learn More
Iron(II) Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10467830

Iron(II) Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. p-7073100 Shop All Fisher Chemical Products
250 g, 250g
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
This item is not returnable. View return policy

Product Code. 10467830

Brand: Fisher Chemical I/1157/50

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7782-63-0
Molecular Formula FeH14O11S
Molecular Weight (g/mol) 278.01
MDL Number MFCD00149719
InChI Key SURQXAFEQWPFPV-UHFFFAOYSA-L
Synonym iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
PubChem CID 62662
ChEBI CHEBI:75836
SMILES O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

Specifications

Melting Point 64°C
Color Blue-Green
pH 3 to 4
Physical Form Solid
Quantity 250 g
IUPAC Name λ²-iron(2+) heptahydrate sulfate
Formula Weight 278.01g/mol
Packaging Plastic powder jar
Vapor Pressure 14.6mmHg at 25°C
compliance-icons
Product Identifier
  • Iron(II) sulfate heptahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Iron (II) Sulfate heptahydrate, Ferroussulfate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Iron(II) Sulfate Heptahydrate, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.