missing translation for 'onlineSavingsMsg'
Learn More

Iron(Ii) Sulfate Heptahydrate, ACS Reagent, Honeywell Fluka™

Catalog Number 15641990
Click to view available options
Quantity:
5 g
250 g
1 kg
6 x 1 kg
This item is not returnable. View return policy
7782-63-0
FeH14O11S
278.01
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
62662
CHEBI:75836
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7782-63-0
FeH14O11S
278.01
MFCD00149719
SURQXAFEQWPFPV-UHFFFAOYSA-L
iron ii sulfate heptahydrate, ferrous sulfate heptahydrate, presfersul, melanterite mineral, iron sulfate heptahydrate, iron 2+ sulfate heptahydrate, fesofor, fesotyme, haemofort, ironate
62662
CHEBI:75836
O.O.O.O.O.O.O.[Fe++].[O-]S([O-])(=O)=O

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 6 x 1 kg
Linear Formula FeSO4 · 7H2O
IUPAC Name iron(2+);sulfate;heptahydrate
Formula Weight 278.01g/mol
Percent Purity ≥99%
Grade ACS Reagent
Vapor Pressure 14.6mmHg (25°C)
Density 1.898g/mL at 25°C
Chemical Name or Material Iron(Ii) Sulfate Heptahydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'