missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron(II) selenide, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Iron(II) selenide is used in electrical semiconductors. It is transformed into superconductor by a purely chemical method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Physical Form | Powder |
Quantity | 10 g |
Assay Percent Range | 99.9% (Metals basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 134.81 |
Odor | Odorless |
Sensitivity | Moisture sensitive |
Density | 6.9g/cm3 |
Chemical Name or Material | Iron(II) selenide |
Safety and Handling
Product Identifier
- Iron(II) selenide
- Danger
- Acute toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'