missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) oxalate dihydrate, 99%, Alfa Aesar™

Brand:  Alfa Aesar A13479.0B

Additional Details : CAS Number : 6047-25-2 Weight : 1.00000kg


Quantity 1000g

 View more versions of this product

Product Code. 11495553

  • 927.00 DKK / 1000g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Applications
It is used as photographic developer for silver bromide-gelatin plates. It imparts greenish-brown tint to optical glass (sunglasses, windshields, railroad car windows), for decorative glassware, pigment for plastics, paints and lacquers. It acts as reagent.

Solubility
Soluble in water. Insoluble in acetic acid.

Notes
Store at room temperature. It is not intended for diagnostic and therapeutic purposes. Avoid contamination with oxidizing agents.
Specifications

Chemical Identifiers

6047-25-2
179.893
NPLZZSLZTJVZSX-UHFFFAOYSA-L
516788
C(=O)(C(=O)[O-])[O-].O.O.[Fe+2]
C2H4FeO6
MFCD00150040
ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™
iron(2+);oxalate;dihydrate

Specifications

Iron(II) oxalate dihydrate
99%
MFCD00150040
Soluble in water. Insoluble in acetic acid.
C(=O)(C(=O)[O-])[O-].O.O.[Fe+2]
179.893
179.90 (143.87 Anhydrous)
2.28
190°C (decomposition)
6047-25-2
C2H4FeO6
ferrous oxalate dihydrate, unii-z6x3ybu50d, z6x3ybu50d, iron ii oxalate hydrate, iron ii oxalate dihydrate, iron oxalate dihydrate, iron 2+ ; oxalate; dihydrate, iron, diaqua ethanedioato 2--kappao1,kappao2-, t-4, iron ii oxalate dihydrate 250g, iron ii oxalate dihydrate, puratronic™
NPLZZSLZTJVZSX-UHFFFAOYSA-L
iron(2+);oxalate;dihydrate
516788
99%
Hygroscopic
1000g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Iron(II) oxalate dihydrate, Puratronic

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P363-Wash contaminated clothing before reuse.
  • P321-Specific treatment (see on this label).
  • P501-Dispose of contents/container to .

SDS Documents