missing translation for 'onlineSavingsMsg'
Learn More

Iron(II) chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4407472
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy
7758-94-3
Cl2Fe
126.75
MFCD00011004
NMCUIPGRVMDVDB-UHFFFAOYSA-L
λ²-iron(2+) dichloride
[Cl-].[Cl-].[Fe++]
This item is not returnable. View return policy
7758-94-3
Cl2Fe
126.75
MFCD00011004
NMCUIPGRVMDVDB-UHFFFAOYSA-L
λ²-iron(2+) dichloride
[Cl-].[Cl-].[Fe++]

Iron(II) chloride is used in the synthesis of iron complexes like iron(II) hydroxide. It acts as a precursor to hydrated iron(III) oxides that are magnetic pigments. It serves as a reducing agent in organic synthesis. Also, used as a reducing flocculating agent in wastewater treatment, especially for wastes containing chromate. It is used as an intermediate, in pigments, processing aids and in solids separation agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1023°C
Melting Point 674°C
Color White
Physical Form Beads
Quantity 1 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,alcohol and acetone. Slightly soluble in benzene. Insoluble in ether.
Formula Weight 126.75
Odor Odorless
Sensitivity Air sensitive; Hygroscopic
Appearance Off-white powder
Vapor Pressure 13.3hPa at 700°C
Density 3.16 g/cm3 at 20°C
Chemical Name or Material Iron(II) chloride, Ultra dry
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Iron(II) chloride, ultra dry
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'