Learn More
Iron(II) chloride tetrahydrate, MilliporeSigma™
Product Code. 15874370
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
Unit Size:
250g
Product Code. Quantity Packaging Unit Size
15874370 250 g Plastic Bottle 250g
1 options
Product Code. 15874370 Supplier Supelco Supplier No. 1038610250
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
For analysis EMSURE™
TRUSTED_SUSTAINABILITY
Specifications
CAS 13478-10-9
Quantity 250 g
Molecular Formula Cl2FeH8O4
Synonym ferrous chloride tetrahydrate, iron ii chloride tetrahydrate, iron dichloride tetrahydrate, cl2fe.4h2o, fecl2.4h2o, iron chloride, tetrahydrate, dichloroiron-water 1/4, iron ii chloride tetra hydrate, ae>> nciu eae r masculineiii
Solubility Information Soluble in water: 1600g/L at 10°C
InChI Key WSSMOXHYUFMBLS-UHFFFAOYSA-L
SMILES O.O.O.O.Cl[Fe]Cl
IUPAC Name dichloroiron;tetrahydrate
Molecular Weight (g/mol) 198.805
PubChem CID 16211588
ChEBI CHEBI:86249
Formula Weight 198.83
Percent Purity ≥98%
Odor Odorless
Packaging Plastic Bottle
Density 1.93g/cm3 at 20°C
Chemical Name or Material Iron(II) chloride tetrahydrate